siRNA Details
| Gene Symbol: EEPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030636.2 | NP_085139.2 | 5, 6 | 1817 | ||
| GenBank mRNA | AB051493.1 | - | 5, 6 | 1558 | ||
| AK027386.1 | - | 5, 6 | 1789 | |||
| BC065518.1 | - | 6, 7 | 1717 | 5755727 | ||

EEPD1
Genomic Map