siRNA Details
Gene Symbol: RNF122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024787.3 | NP_079063.2 | 6 | 1592 | ||
XM_017013846.1 | XP_016869335.1 | 6 | 1488 | |||
GenBank mRNA | AK022588.1 | - | 6 | 1589 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024787.3 | NP_079063.2 | 6 | 1592 | ||
XM_017013846.1 | XP_016869335.1 | 6 | 1488 | |||
GenBank mRNA | AK022588.1 | - | 6 | 1589 |