siRNA Details
| Gene Symbol: Gins3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030198.3 | NP_084474.1 | 3 | 2014 | ||
| GenBank mRNA | AK019225.1 | - | 3 | 2015 | ||
| AK146574.1 | - | 3 | 2046 | |||
| AK160606.1 | - | 3 | 2015 | |||
| BC064746.1 | - | 3 | 2003 | 30095310 | ||

Gins3
Genomic Map