siRNA Details
| Gene Symbol: Slc46a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026740.2 | NP_081016.2 | 5 | 1420 | ||
| GenBank mRNA | AK003278.1 | - | 5 | 1403 | ||
| AK170505.1 | - | 5 | 1413 | |||
| AK220201.1 | - | 5 | 1380 | |||
| BC024522.1 | - | 4 | 556 | |||
| BC057976.1 | - | 5 | 1405 | 5052851 | ||

Slc46a1
Genomic Map