siRNA Details
| Gene Symbol: Ripk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139342.1 | NP_647558.1 | 6, 7 | 1068 | ||
| XM_006251928.2 | XP_006251990.1 | 5, 6 | 1920 | |||
| XM_008770661.1 | XP_008768883.1 | 4, 5 | 881 | |||
| XM_008770662.1 | XP_008768884.1 | 5, 6 | 1252 | |||
| GenBank mRNA | AF036537.2 | - | 6, 7 | 1068 | ||
| BC158580.1 | - | 4, 5 | 635 | 7378607 | ||

Ripk3
Genomic Map