siRNA Details
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009455.1 | NP_001009455.1 | 3 | 1319 | ||
GenBank mRNA | BC087086.1 | - | 3 | 1319 | 7189809 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009455.1 | NP_001009455.1 | 3 | 1319 | ||
GenBank mRNA | BC087086.1 | - | 3 | 1319 | 7189809 |