siRNA Details
| Gene Symbol: Chtop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014175.1 | NP_001014197.1 | 5 | 1139 | ||
| XM_006232710.2 | XP_006232772.1 | 6 | 2107 | |||
| XM_006232711.2 | XP_006232773.1 | 6 | 2023 | |||
| XM_006232713.2 | XP_006232775.1 | 6 | 2019 | |||
| XM_006232715.2 | XP_006232777.1 | 5 | 1885 | |||
| GenBank mRNA | BC081985.1 | - | 5 | 1139 | 7128951 | |
| BC100084.1 | - | 6 | 1055 | 7385093 | ||

Chtop
Genomic Map