siRNA Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 8 | 1037 | |||
| NR_024429.1 | 8 | 1133 | ||||
| NR_024431.2 | 10 | 1329 | ||||
| GenBank mRNA | AK097923.1 | - | 4 | 1748 | ||
| AK098249.1 | - | 10 | 1774 | |||
| BC102000.1 | - | 7 | 880 | |||
| BC102001.1 | - | 7 | 880 | |||
| BC105703.1 | - | 7 | 976 | |||

ZMIZ1-AS1
Genomic Map