siRNA Details
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149372.1 | - | 6 | 735 | ||
BC125617.1 | - | 6 | 733 | |||
BC132393.1 | - | 6 | 733 | |||
M23109.1 | - | 6 | 698 | |||
M26236.1 | - | 1 | 197 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149372.1 | - | 6 | 735 | ||
BC125617.1 | - | 6 | 733 | |||
BC132393.1 | - | 6 | 733 | |||
M23109.1 | - | 6 | 698 | |||
M26236.1 | - | 1 | 197 |