siRNA Details
| Gene Symbol: Epha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023580.4 | NP_076069.2 | 12, 13 | 2137 | ||
| GenBank mRNA | AF131197.1 | - | 12, 13 | 2079 | ||
| AK028478.1 | - | 12, 13 | 2138 | |||
| BC071215.1 | - | 12, 13 | 2101 | 4196138 | ||
| U18084.1 | - | 8, 9 | 1240 | |||

Epha1
Genomic Map