siRNA Details
| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171141.1 | - | 5 | 762 | ||
| BC020067.1 | - | 5 | 749 | 3991510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caap1| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171141.1 | - | 5 | 762 | ||
| BC020067.1 | - | 5 | 749 | 3991510 | ||