siRNA Details
| Gene Symbol: Tmem86a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026436.3 | NP_080712.1 | 3 | 1124 | ||
| GenBank mRNA | AK007864.1 | - | 3 | 1125 | ||
| AK075589.1 | - | 3 | 1098 | |||
| AK213808.1 | - | 1 | 101 | |||
| BC056619.1 | - | 3 | 1091 | 4987281 | ||

Tmem86a
Genomic Map