siRNA Details
| Gene Symbol: Memo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133771.2 | NP_598532.1 | 5 | 468 | ||
| GenBank mRNA | AF363447.1 | - | 6 | 530 | ||
| AK159560.1 | - | 5 | 432 | |||
| BC013819.1 | - | 5 | 485 | 3489363 | ||
| BF466279.1 | - | 5 | 468 | |||

Memo1
Genomic Map