siRNA Details
Gene Symbol: TOR1AIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199260.1 | NP_001186189.1 | 7 | 2040 | ||
NM_145034.4 | NP_659471.1 | 6 | 1621 | |||
XM_005244939.4 | XP_005244996.1 | 7 | 2013 | |||
XM_017000474.1 | XP_016855963.1 | 8 | 2158 | |||
XM_017000475.1 | XP_016855964.1 | 6 | 1882 | |||
XM_017000476.1 | XP_016855965.1 | 3 | 1322 | |||
GenBank mRNA | AF464140.1 | - | 6 | 1615 | ||
BC101532.1 | - | 4 | 1325 | 8069038 | ||
BC112225.1 | - | 4 | 1325 | 8327693 |
