siRNA Details
Gene Symbol: DANCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024031.1 | 2 | 161 | |||
GenBank mRNA | BC015222.1 | - | 1 | 99 | ||
BC045566.1 | - | 2 | 110 | |||
D28589.1 | - | 1 | 98 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DANCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024031.1 | 2 | 161 | |||
GenBank mRNA | BC015222.1 | - | 1 | 99 | ||
BC045566.1 | - | 2 | 110 | |||
D28589.1 | - | 1 | 98 |