siRNA Details
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004354.2 | NP_001004354.1 | 1 | 617 | ||
GenBank mRNA | BC053618.1 | - | 1 | 617 | 6143133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004354.2 | NP_001004354.1 | 1 | 617 | ||
GenBank mRNA | BC053618.1 | - | 1 | 617 | 6143133 |