siRNA Details
Gene Symbol: FAM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100411.2 | NP_001093881.1 | 17 | 3211 | ||
NM_001288576.1 | NP_001275505.1 | 16 | 3106 | |||
NM_024581.5 | NP_078857.5 | 18 | 3749 | |||
GenBank mRNA | AJ420503.1 | - | 2 | 2229 | ||
AJ420563.1 | - | 1 | 1136 | |||
AK024004.1 | - | 11 | 1553 | |||
AK091682.1 | - | 16 | 3102 | |||
AK127744.1 | - | 14 | 2051 | |||
AL136767.1 | - | 8 | 959 | |||
AU160036.1 | - | 1 | 293 | |||
AY131226.1 | - | 14 | 2025 | |||
BC009055.1 | - | 10 | 1341 | |||
BC060769.1 | - | 18 | 3749 | 30337859 | ||
BX538186.1 | - | 17 | 3191 |
