siRNA Details
Gene Symbol: Gucy1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204340.1 | NP_001191269.1 | 10 | 1261 | ||
NM_172810.3 | NP_766398.1 | 10 | 1306 | |||
GenBank mRNA | AF038500.1 | - | 4 | 460 | ||
AK047129.1 | - | 10 | 1307 | |||
AY236513.1 | - | 8 | 980 | |||
BC137868.1 | - | 10 | 1258 | 8860887 | ||
BC144988.1 | - | 10 | 1255 | 9053536 |
