siRNA Details
Gene Symbol: Olfr692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146355.1 | NP_666467.1 | 1 | 259 | ||
GenBank mRNA | BC119096.1 | - | 1 | 357 | 8733845 | |
BC120788.1 | - | 1 | 357 | 40129711 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146355.1 | NP_666467.1 | 1 | 259 | ||
GenBank mRNA | BC119096.1 | - | 1 | 357 | 8733845 | |
BC120788.1 | - | 1 | 357 | 40129711 |