siRNA Details
Gene Symbol: B3gnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081167.1 | NP_001074636.1 | 2 | 772 | ||
GenBank mRNA | AK140429.1 | - | 1 | 346 | ||
BC039789.1 | - | 2 | 788 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B3gnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081167.1 | NP_001074636.1 | 2 | 772 | ||
GenBank mRNA | AK140429.1 | - | 1 | 346 | ||
BC039789.1 | - | 2 | 788 |