siRNA Details
Gene Symbol: Evx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007966.4 | NP_031992.1 | 1 | 363 | ||
GenBank mRNA | BC126907.1 | - | 1 | 121 | 40059181 | |
X54239.1 | - | 1 | 364 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Evx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007966.4 | NP_031992.1 | 1 | 363 | ||
GenBank mRNA | BC126907.1 | - | 1 | 121 | 40059181 | |
X54239.1 | - | 1 | 364 |