siRNA Details
| Gene Symbol: Zbbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172515.2 | NP_766103.2 | 15, 16 | 2106 | ||
| GenBank mRNA | AK029899.1 | - | 15, 16 | 2099 | ||
| AK133102.1 | - | 15, 16 | 2157 | |||
| BC050789.1 | - | 15, 16 | 2106 | 6706914 | ||

Zbbx
Genomic Map