siRNA Details
| Gene Symbol: Tbce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082940.1 | - | 9 | 949 | ||
| AK167383.1 | - | 9 | 937 | |||
| AK167848.1 | - | 9 | 933 | |||
| AY082332.1 | - | 9 | 903 | |||
| BC050206.1 | - | 9 | 911 | 30009463 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Tbce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082940.1 | - | 9 | 949 | ||
| AK167383.1 | - | 9 | 937 | |||
| AK167848.1 | - | 9 | 933 | |||
| AY082332.1 | - | 9 | 903 | |||
| BC050206.1 | - | 9 | 911 | 30009463 | ||
Tbce
Genomic Map