siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 4, 5 | 459 | ||
| GenBank mRNA | AK002768.1 | - | 4, 5 | 361 | ||
| AK016995.1 | - | 3, 4 | 929 | |||
| BC022718.1 | - | 3, 4 | 433 | |||
| BC022733.1 | - | 4, 5 | 457 | 4506628 | ||
| BG297132.1 | - | 4, 5 | 456 | |||
| DT911344.1 | - | 4, 5 | 433 | |||

Ccdc163
Genomic Map