siRNA Details
| Gene Symbol: Mybpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252372.1 | NP_001239301.1 | 26 | 3519 | ||
| NM_175418.5 | NP_780627.2 | 27 | 3561 | |||
| GenBank mRNA | AK078761.1 | - | 7 | 955 | ||
| AK132188.1 | - | 26 | 3392 | |||
| BB669565.1 | - | 1 | 50 | |||
| BC037516.1 | - | 1 | 285 | |||
| BC062158.1 | - | 27 | 3423 | 30134842 | ||

Mybpc1
Genomic Map