siRNA Details
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011785.3 | NP_035915.3 | 8 | 797 | ||
GenBank mRNA | AF124142.1 | - | 8 | 787 | ||
BC066861.1 | - | 8 | 797 | 30089997 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011785.3 | NP_035915.3 | 8 | 797 | ||
GenBank mRNA | AF124142.1 | - | 8 | 787 | ||
BC066861.1 | - | 8 | 797 | 30089997 |