siRNA Details
Gene Symbol: D430041D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033347.2 | NP_001028519.2 | 19 | 3961 | ||
GenBank mRNA | AK036073.1 | - | 2 | 1765 | ||
AK052518.1 | - | 10 | 1864 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D430041D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033347.2 | NP_001028519.2 | 19 | 3961 | ||
GenBank mRNA | AK036073.1 | - | 2 | 1765 | ||
AK052518.1 | - | 10 | 1864 |