siRNA Details
Gene Symbol: Olfr1367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146533.1 | NP_666744.1 | 1 | 912 | ||
GenBank mRNA | BC138110.1 | - | 1 | 932 | 8861129 | |
BC138111.1 | - | 1 | 932 | 8861130 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146533.1 | NP_666744.1 | 1 | 912 | ||
GenBank mRNA | BC138110.1 | - | 1 | 932 | 8861129 | |
BC138111.1 | - | 1 | 932 | 8861130 |