siRNA Details
Gene Symbol: Hdgfrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013886.4 | NP_038914.2 | 3 | 724 | ||
GenBank mRNA | AB029493.1 | - | 3 | 287 | ||
AK019487.1 | - | 3 | 387 | |||
AK044858.1 | - | 3 | 549 | |||
AK045542.1 | - | 3 | 724 | |||
AK051401.1 | - | 3 | 646 | |||
AK162418.1 | - | 3 | 367 | |||
AK162967.1 | - | 3 | 288 | |||
BC055734.1 | - | 3 | 311 | 6810355 |
