siRNA Details
Gene Symbol: Nos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021838.2 | NP_068610.1 | 6, 7 | 817 | ||
XM_006235872.1 | XP_006235934.1 | 6, 7 | 846 | |||
GenBank mRNA | AB176831.1 | - | 6, 7 | 817 | ||
AF085195.1 | - | 6, 7 | 818 | |||
AY695391.1 | - | 6, 7 | 772 |
