siRNA Details
Gene Symbol: Cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138877.1 | NP_620232.1 | 5 | 402 | ||
XM_006242065.2 | XP_006242127.1 | 5 | 430 | |||
GenBank mRNA | BC062066.1 | - | 5 | 418 | 5599255 | |
D00636.1 | - | 5 | 402 | |||
J03867.1 | - | 5 | 397 |
