siRNA Details
| Gene Symbol: Myh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013194.1 | NP_037326.1 | 5056 | |||
| GenBank mRNA | U31463.1 | - | 5056 | |||
| Gene Symbol: LOC100911597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750378.3 | XP_003750426.1 | 35 | 5268 | ||

Myh9
Genomic Map