siRNA Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105968.1 | NP_001099438.1 | 13 | 1684 | ||
| XM_006250240.2 | XP_006250302.1 | 12 | 2585 | |||
| XM_006250241.1 | XP_006250303.1 | 13 | 1749 | |||
| XM_006250243.1 | XP_006250305.1 | 12 | 1392 | |||

Ppox
Genomic Map