siRNA Details
Gene Symbol: LOC102552318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760184.1 | XP_008758406.1 | 7 | 995 | ||
XM_008760185.1 | XP_008758407.1 | 7 | 994 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102552318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760184.1 | XP_008758406.1 | 7 | 995 | ||
XM_008760185.1 | XP_008758407.1 | 7 | 994 |