siRNA Details
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005540.1 | NP_001005540.1 | 8, 9 | 1213 | ||
XM_008761380.1 | XP_008759602.1 | 9, 10 | 1711 | |||
GenBank mRNA | BC082053.1 | - | 8, 9 | 1213 | 7132147 |
