siRNA Details
| Gene Symbol: Zfp672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007669.1 | NP_001007670.1 | 3 | 1230 | ||
| XM_006246389.2 | XP_006246451.1 | 3 | 1500 | |||
| XM_006246390.2 | XP_006246452.1 | 3 | 1128 | |||
| XM_006246391.2 | XP_006246453.1 | 3 | 1323 | |||
| XM_008767686.1 | XP_008765908.1 | 2 | 6262 | |||
| XM_008767687.1 | XP_008765909.1 | 3 | 1447 | |||
| GenBank mRNA | BC081919.1 | - | 3 | 1230 | 7112624 | |

Zfp672
Genomic Map