siRNA Details
Gene Symbol: Csmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037327.1 | NP_001032404.1 | 69 | 10628 | ||
GenBank mRNA | DQ124115.1 | - | 69 | 10628 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037327.1 | NP_001032404.1 | 69 | 10628 | ||
GenBank mRNA | DQ124115.1 | - | 69 | 10628 |