siRNA Details
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198766.1 | NP_942061.1 | 3 | 272 | ||
XM_006227783.2 | XP_006227845.1 | 3 | 287 | |||
GenBank mRNA | BC058157.1 | - | 3 | 272 | 6890640 |
