siRNA Details
Gene Symbol: Actl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024788.1 | NP_001019959.1 | 1 | 2428 | ||
GenBank mRNA | AY635905.1 | - | 1 | 2428 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Actl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024788.1 | NP_001019959.1 | 1 | 2428 | ||
GenBank mRNA | AY635905.1 | - | 1 | 2428 |