siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012275.2 | NP_001012275.1 | 7 | 799 | ||
| XM_006243784.2 | XP_006243846.1 | 7 | 1267 | |||
| XM_006243785.2 | XP_006243847.1 | 7 | 1264 | |||
| XM_006243787.2 | XP_006243849.1 | 6 | 488 | |||
| XM_008766537.1 | XP_008764759.1 | 7 | 952 | |||
| XM_008766538.1 | XP_008764760.1 | 7 | 955 | |||
| GenBank mRNA | CV126331.1 | - | 3 | 303 | ||

Arih2
Genomic Map