siRNA Details
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031685.2 | NP_113873.1 | 6 | 579 | ||
GenBank mRNA | BC061994.1 | - | 6 | 579 | 5599557 | |
U91539.1 | - | 6 | 578 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031685.2 | NP_113873.1 | 6 | 579 | ||
GenBank mRNA | BC061994.1 | - | 6 | 579 | 5599557 | |
U91539.1 | - | 6 | 578 |