siRNA Details
Gene Symbol: Mmp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031757.1 | NP_113945.1 | 7 | 1175 | ||
GenBank mRNA | AB023659.1 | - | 7 | 1175 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mmp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031757.1 | NP_113945.1 | 7 | 1175 | ||
GenBank mRNA | AB023659.1 | - | 7 | 1175 |