siRNA Details
Gene Symbol: Ptger4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032076.3 | NP_114465.3 | 2 | 1293 | ||
XM_006231996.2 | XP_006232058.1 | 3 | 3862 | |||
GenBank mRNA | D28860.1 | - | 3 | 1473 | ||
U94709.1 | - | 2 | 1293 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
