siRNA Details
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031515.3 | NP_113703.1 | 2 | 153 | ||
XM_008763352.1 | XP_008761574.1 | 2 | 2746 | |||
XM_008763353.1 | XP_008761575.1 | 3 | 190 | |||
XM_008763354.1 | XP_008761576.1 | 3 | 335 | |||
XM_008763355.1 | XP_008761577.1 | 2 | 2746 | |||
GenBank mRNA | BC126086.1 | - | 3 | 291 | 8362025 | |
FQ230946.1 | - | 3 | 334 | |||
U09793.1 | - | 2 | 153 |
