siRNA Details
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025632.2 | NP_001020803.2 | 12 | 1255 | ||
XM_006251201.2 | XP_006251263.2 | 12 | 1240 | |||
XM_006251202.2 | XP_006251264.2 | 12 | 1237 | |||
XM_006251203.2 | XP_006251265.2 | 12 | 1266 | |||
GenBank mRNA | BC098822.1 | - | 3 | 289 | ||
BC162004.1 | - | 12 | 1255 | 9089248 | ||
FQ221459.1 | - | 3 | 402 |
Gene Symbol: LOC100912481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749683.2 | XP_003749731.1 | 12 | 1260 | ||
XM_003749684.2 | XP_003749732.1 | 13 | 1275 | |||
XM_006236016.1 | XP_006236078.1 | 13 | 1278 | |||
XM_006236017.1 | XP_006236079.1 | 12 | 1257 |
