siRNA Details
Gene Symbol: Ccdc101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114502.1 | NP_001107974.1 | 7, 8 | 763 | ||
XM_006230231.2 | XP_006230293.1 | 7, 8 | 869 | |||
XM_006230232.2 | XP_006230294.1 | 7, 8 | 773 | |||
GenBank mRNA | FQ210727.1 | - | 7, 8 | 738 | ||
FQ224509.1 | - | 7, 8 | 738 | |||
FQ232716.1 | - | 7, 8 | 738 |
