siRNA Details
| Gene Symbol: Olr495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000310.1 | NP_001000310.1 | 1 | 892 | ||
| Gene Symbol: LOC100909940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749508.2 | XP_003749556.1 | 1 | 892 | ||

Olr495
Genomic Map