siRNA Details
| Gene Symbol: Rec114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106825.1 | NP_001100295.1 | 5 | 654 | ||
| XM_006243157.2 | XP_006243219.1 | 5 | 786 | |||
| XM_006243158.2 | XP_006243220.1 | 5 | 799 | |||
| XM_006243160.2 | XP_006243222.1 | 5 | 802 | |||

Rec114
Genomic Map