siRNA Details
Gene Symbol: LOC102546596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257622.2 | XP_006257684.1 | 2 | 388 |
Gene Symbol: LOC102554207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240812.2 | XP_006240874.1 | 2 | 388 |
