siRNA Details
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039017.1 | NP_001034106.1 | 3 | 273 | ||
XM_006246502.2 | XP_006246564.1 | 3 | 332 | |||
XM_008767821.1 | XP_008766043.1 | 3 | 311 | |||
XM_008767822.1 | XP_008766044.1 | 3 | 616 | |||
XM_008767823.1 | XP_008766045.1 | 3 | 619 | |||
GenBank mRNA | BC099105.1 | - | 3 | 273 | 7460369 |
